Weller® Cordless Soldering Iron Kit, Cooper Tools, Apex
Supplier: Cooper Tools
PSI100K
37698-362EA 378.81 USD
37698-362
Weller® Cordless Soldering Iron Kit, Cooper Tools, Apex
Soldering Supplies Soldering Irons
Professional-quality Portasol® heat tool to solder, heat shrink, and cut rope
Butane fuel level is visible through a window on the body of the tool. Built-in fuel filter reduces fuel contamination. Cap includes auto safety shut-off switch. Operates up to two hours per refill. Temperature is adjustable up to 580°C (1076°F). Kit contains iron, sponge, stand, and extra tips, all packed in a plastic case.
Order Now
SPECIFICATIONS
Environmentally Preferable