LPG Sample Cylinders, Koehler
Supplier: Koehler
K27851 K27852 K27853 K27854 K27856-00000
37000-216EA 3097.67 USD
37000-216 37000-218 37000-220 37000-222 37000-224
LPG Sample Cylinders, Koehler
Samplers Air Samplers
Welded stainless steel cylinders obtain representative samples of liquefied petroleum gases (LPG)
Two-valve type instruments feature a 20% outage tube and built-in pressure relief valve preset between 540 and 600psi (38 and 42kg/cm2).
Certificaciones: Conform to specifications: ASTM D1265, GPA 2140.