Sample Thieves (Bacon Bombs), Koehler
Supplier: Koehler
K27772 K27701 K27760 K27761 K27762 K27770 K27771 K27781 K27782 K27785 K27791 K27792 K27795
37000-202EA 1450.73 USD
37000-202 37000-188 37000-192 37000-194 37000-196 37000-198 37000-200 37000-204 37000-206 37000-208 37000-210 37000-212 37000-214
Sample Thieves (Bacon Bombs), Koehler
Samplers Fluid Samplers
Devices obtain samples from any level in storage tanks, tank cars, and drums
A plunger assembly opens to admit the sample when the thief strikes the bottom of the tank or is triggered by a secondary trip line. Plunger locking cam provides a tight closure during transport, except for 2.8cm (11/8") diameter models, which are designed for sampling through narrow pipes and openings.
Order Now
SPECIFICATIONS
Environmentally Preferable