High Temperature Dropping Point Apparatus, Koehler
Supplier: Koehler
K19400 K19410
37000-128EA 13286.26 USD
37000-128 37000-130
High Temperature Dropping Point Apparatus, Koehler
Petroleum Testing Apparatus
The high temperature dropping point test permits higher temperatures than the ASTM D566 method and uses a different heating procedure.
  • Six-sample testing capability
  • Microprocessor programmable high accuracy temperature control
The test cell is inserted in an aluminum block oven maintained at a constant temperature that is higher than the expected dropping point of the sample. The sample temperature then rises to the dropping point without operator control.
Tests dropping points of lubricating greases at temperatures of up to 400 °C. Dual LEDMore Product Information