Anti-CD63 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H5C6]
Supplier: Biolegend
353005 353006
353005-BLEA 110.4 USD
353005-BL 353006-BL
Anti-CD63 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H5C6]
Antibodies
FITC anti-human CD63 [H5C6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/More Product Information