VORTEX 3 Orbital Shakers, 230 V
Supplier: IKA Works
3340000
33993-876EA 981.68 USD
33993-876
VORTEX 3 Orbital Shakers, 230 V
Shakers and Mixers Orbital Shakers
Orbital shakers are suitable for touch-operated or short-time continuous operation
  • 230 V model
Shaker attachments and inserts securely click onto units in any position and allow various applications to be performed. The one-hand attachment is 88 mm (3⁷/₁₆") in diameter for larger vessels. The universal attachment allows use of various inserts for tubes, flasks, and plates.
Self-ventilating motor is encased in a sturdy zinc die-cast housing. Compact shakers feature wide, infinitely adjustable speed ranges. Special silicone base feet with ultra-highMore Product Information