Hemispherical Mantles, Series O, Glas-Col®
Supplier: Glas-Col
100AO394 100AO398 100AO400 100AO404 0616700220 0616700229 100AO406 100AO408 100AO410 100AO412 100AO414 100AO416 100AO396 100AO402
33787-028EA 314.53 USD
33787-028 33787-061 33787-083 33787-120 33787-130 33787-132 33787-134 33787-136 33787-141 33787-163 33787-185 33787-200 33787-221 33787-265 33787-040 33787-108
Hemispherical Mantles, Series O, Glas-Col®
Heaters Heating Mantles
These hemispherical-style mantles cover only the bottom half of the flask, allowing full view of contents
Constructed of flexible fabrics for effective heating and protection of glass vessels. Choose from glass fabric exterior or silicone-impregnated glass fabric exterior. Mantles accommodate single and multi-neck flasks and adapt easily to unusual shapes. Withstand 450°C (840°F) internal operating temperature.
Información para pedidos: The use of an extension support is required (see 33793-018 series). Mantles include a separable 1.2m (4') long two-wire cord and locking connector.More Product Information