Stir Mantles, Large Capacity, Glas-Col®
Supplier: Glas-Col
100DEMS116 100DEMS117 100DEMS119 100DEMS120 100DEMS121 100DEMS122
33725-070EA 3458.89 USD
33725-070 33725-072 33725-074 33725-076 33725-078 33725-080 33725-082
Stir Mantles, Large Capacity, Glas-Col®
Heaters Heating Mantles
Heating and stirring in one unit.
  • 1/50 HP motor
  • Variable speed control
  • Large capacity
  • Total mixing and heating management
  • For use with spherical flask
This system has the power to mix larger volumes in a flask without an overhead stirrer. Powerful motor/magnet combination generates speed range of 100-1850 rpm. Variable motor speed control has detachable cord. Mantle is aluminum housed with fabric interior to softly nestle glassware. Ideal for temperatures up to 400 degrees C. The mantle is mounted on 4" feet to allow bench clearance for the stir motorMore Product Information