SterileSorb™ Wipers, Contec®
Supplier: Contec
C2-1212IR C2-99IR/25
33502-292CS 723.07 USD
33502-292 68310-099
SterileSorb™ Wipers, Contec®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
SterileSorb™ Wipes are a hydroentangled blend of 55% cellulose and 45% polyester. They are highly sorbent with excellent durability and good cleanliness. SterileSorb™ wipes are strong and durable wet or dry, making them excellent for use in general cleaning and spill pick up.
  • Wipes provide low levels of particles and extractables
  • An excellent cost effective, general-purpose wipe
  • Easy to open, even when wearing gloves
These wipes are packaged in sterile peelable outer bags and linear tear inner bags specifically designed for sterile products. EmbossedMore Product Information