Supreno® SE Powder-Free Nitrile Gloves, Microflex®, Ansell
Supplier: ANSELL COMMERCIAL MEXICO SA DE MX
SU-690-M SU-690-XL SU-690-XS SU-690-S SU-690-L
32916-662CS 477.23 USD
32916-662 32916-666 32916-668 32916-660 32916-664
Supreno® SE Powder-Free Nitrile Gloves, Microflex®, Ansell
Gloves
Supreno® powder-free gloves are made of soft, durable nitrile formulated for improved strength and comfort.
  • Soft, powder-free
  • Formulated for improved strength and comfort
  • Thickness: 7.9 mil (finger)
Interior polymer coating makes gloves easy to don. Textured fingers allow for reliable grip in wet and dry conditions. Online chlorination.
Información de suministro: Gloves are packaged 100 gloves per dispenser box.
Precaución: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.