Stir Bar, Micro, PTFE, Dynalon
Supplier: Dynalon
303495 303505 303475
30620-420CS 128.81 USD
30620-420 30620-422 30621-014
Stir Bar, Micro, PTFE, Dynalon
Stirrers Magnetic Stirring Bars
Designed for stirring in test tubes, micro-beakers, and similar vessels
Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance and is autoclavable. Always store in pairs to maintain magnetic strength.