Drying Rack, High Impact Polystyrene, Dynalon
Supplier: Dynalon
259184
30620-320EA 444.91 USD
30620-320
Drying Rack, High Impact Polystyrene, Dynalon
Racks Draining Racks
This 72 place drying rack is molded of high impact polystyrene
It comes supplied with 72 large pegs, draining tubes, fastening stoppers and expansion hanging hooks which allow coupling with additional racks. The pegs are 4" long and the drip ledge measures 3". Expandable fasteners to connect additional racks included and interchangeable removable pegs