FluoroTrans® PVDF Transfer Membranes, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
PVM020C-099 BSP0157 PVM020C-160 BSP0159 BSP0161 BSP0158 PVM020C-195 PVM020C-196
29301-808EA 626.21 USD
29301-808 29301-850 29301-862 29301-854 29301-856 29301-852 29301-864 29301-866
FluoroTrans® PVDF Transfer Membranes, Cytiva (Formerly Pall Lab)
Transfer Membranes
Naturally hydrophobic PVDF, ideal for a wide variety of protein-analysis applications. The family of FluroTrans® media are white, microporous solid phase supports that bind proteins tenaciously via hydrophobic interactions. FluoroTrans® membrane is optimised for N-terminal protein sequencing. FluoroTrans® W membranes are optimised for Western transfer applications. They allow for sensitive protein detection with low background and very low protein burn-through. Immobilised proteins can be visually detected with Coomassie® Blue, Amido black, Ponceau S, and colloidal gold.
  • High protein
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable