Corning® Filter Top Tubes, Sterile, Corning
Supplier: Corning
430320 430314
28680-120CS 100.54 USD
28680-120 28680-121
Corning® Filter Top Tubes, Sterile, Corning
Filters Bottle Top Filters
Cellulose acetate membrane, PS funnel and PP collection tube.
  • 50 mm diameter membrane
  • Minimises unnecessary transfers by filtering directly into 50 ml centrifuge tube
  • Includes two centrifuge tube stands with each case
  • Each PP centrifuge tube is supplied with an individually wrapped cap for storage
  • Individually packaged, sterile, certified non pyrogenic