Whatman™ Kjeldahl Analysis Weighing Paper, Whatman products (Cytiva)
Supplier: Cytiva
10347670 10347671 10347672 10347673
28498-000PK 214.98 USD
28498-000 28498-002 28498-004 28498-006
Whatman™ Kjeldahl Analysis Weighing Paper, Whatman products (Cytiva)
Weighing Papers
Grade B-2 Whatman weighing paper from Cytiva dissolves residue-free in acid solution, which is critical in applications such as Kjeldahl analysis of nitrogen in soil.
  • Thin (0.04 mm) and lightweight (43 g/m²)
  • Transparent and smooth parchment surface
  • Simple weighing out and sample transfer
  • Reliable for quantitative transfer
  • Range of sheet sizes for ease-of-use
  • Low nitrogen paper that is residue-free for accurate chemical analysis
Cytiva also offers weighing boats for Kjeldahl analysis.
Packs of 500 Grade B-2 Whatman weighing paper are available inMore Product Information