Cliniplate™ 384-Well Microplates, Thermo Scientific
Supplier: Thermo Fisher Scientific
95040000
28298-614CS 891.49 USD
28298-614
Cliniplate™ 384-Well Microplates, Thermo Scientific
Microplates HTS Microplates
These polystyrene microplates are designed for high throughput screening and are ideal for a wide range of measurement techniques
Clear plates are ideal for photometry applications. White and black plates are designed for assays utilizing fluorescent or luminescent markers. The 384-well format enables higher sample density, lower consumption of reagents, and reduced overall costs. Plates are designed for use with instrumentation adapted for 384-well plates. Cliniplates™ also feature excellent optical performance and rounded wells for improved mixing.
Información para pedidos: SterileMore Product Information