Owl™ Sequencing Gel Casters, Model SGC, Thermo Scientific
Supplier: Thermo Fisher Scientific
SGC-2
27372-830EA 705.94 USD
27372-830 27372-832
Owl™ Sequencing Gel Casters, Model SGC, Thermo Scientific
Blotting Kits
Cast bubble-free sequencing gels in under three minutes.
  • Capillary action draws gel between the plates
  • Surface tension eliminates leaks
  • No tape or special assembly is needed
Easy-to-use gel casters are adjustable from 20 cm (77/8") wide to 42 cm (161/2") wide.
Información para pedidos: Systems include adjustable casting platform, width adjustment knobs, and 12 tempered steel binder clamps.
Order Now
SPECIFICATIONS
Environmentally Preferable