CentriVap® Centrifugal Concentration System Accessories, Labconco®
Supplier: Labconco
7814300 7991000 7814800 7814900 7815000 7815200 7815300 7464300 7461900 7460900 7456600P 7450700 7462900 7455100 7455200 7474000 7995600 7397605 7397700 7450701 7455001 7455101 7455201 7461901 7462901 7814500 7455000 7553100 7824700 7824702 7553500 7553501
15398-028EA 167 USD
15398-028 26617-040 26675-878 26675-879 26675-881 26675-884 26675-885 26676-010 37000-590 37000-596 37001-388 37001-390 37001-392 37001-394 37001-398 37001-400 74920-006 83009-292 89004-964 89004-966 89041-252 89041-254 89041-256 89041-258 89041-260 89041-262 26675-877 37001-396 89239-682 89259-754 89259-756 75779-942 75779-944
CentriVap® Centrifugal Concentration System Accessories, Labconco®
Concentrators
For use with all CentriVap® centrifugal concentration systems, except DNA system.
CentriZap® strobe light enables user to shine a strobe light into the chamber to easily view samples while the rotor is spinning. Samples appear to be standing still. Includes a holster bracket (with mounting hardware) that attaches to the right side of the unit. Glass lid provides additional protection from solvents and corrosives that attack acrylic. Vacuum controller provides manual control and monitoring of vacuum levels. Analog gauge shows vacuum from 0–762 mm Hg (0–30" Hg). Three-way valve isMore Product Information