Barnstead Bantam™ Deionizer Holders, Thermo Scientific
Supplier: Thermo Fisher Scientific
D0800 D0805
26302-082EA 1593.1 USD
26302-082 26302-083
Barnstead Bantam™ Deionizer Holders, Thermo Scientific
Water Purification Systems Deionizers
These holders connects directly to raw water line and delivers demineralized water in continuous flow at atmospheric pressure.
  • Simple thumbwheel arrangement permits cartridge change without tools
  • Direct reading meter indicates purity of effluent at all times
  • Pedestal is removable if wall-mounting is preferred
  • Shows when cartridge needs replacing
Meter calibrated at 0-19 ppm (as NaCl) and in ohms resistance from 25000 to 18000000 ohms-cm.
Información de suministro: Supplied with 0.9 m (3') plastic tube to connect to water line, inlet flow adjustmentMore Product Information