VWR® Cork Sheet
Supplier: VWR International
23420-708
23420-708PK 91.8 USD
23420-708
VWR® Cork Sheet
Surface Protectors Bench Protectors
Use to protect workbenchtop from heat, acids, and alkalies.
  • Protects against heat, acids, and alkalies
Cork sheet can also be used as a dissecting pad.