Bentron™ Conductive Tote Boxes, Metro International
Supplier: Metro
TB93120CAS TB92050CAS TB93060CAS TB92080CAS TB91060CAS TB93080CAS TB93050CAS TB93030CAS TB92060CAS TB92035CAS TB91035CAS
22221-242EA 227.88 USD
22221-242 22221-292 22221-246 22221-288 22221-296 22221-244 22221-284 22221-286 22221-290 22221-294 22221-298
Bentron™ Conductive Tote Boxes, Metro International
Boxes ESD-Safe Boxes
Tote boxes offer safe and efficient storage of items in bulk or in compartmentalized fashion with the use of optional dividers and covers
These conductive tote boxes are made of polypropylene.
Boxes are durable, stackable, and feature high impact strength. There is a smooth area on each side of the box that is a convenient place to affix labels and bar codes. ESD Classification: Conductive.
Información para pedidos: Totes are also available in natural gray and blue static dissipative Benstat™. Contact your Avantor representative for more information.
Order Now
SPECIFICATIONS
Environmentally Preferable