Isonicotinonitrile

Warning

Synonyms: 4-Pyridinecarbonitrile, Pyridine-4-carbonitrile, 4-Cyanopyridine

C95005-100G
220116-845EA 25.52 USD
220116-845
Isonicotinonitrile
Isonicotinonitrile
Formula: C₆H₄N₂
MW: 104.11 g/mol
Boiling Pt: 195.4 °C (1013 hPa)
Melting Pt: 78 - 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00006417
CAS Number: 100-48-1
EINECS: 202-856-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR