2,5-Dibromo-4-picoline

Synonyms: 2,5-Dibromo-4-methylpyridine

708143-1G
220092-500EA 69.68 USD
220092-500
2,5-Dibromo-4-picoline
2,5-Dibromo-4-picoline
Formula: C₆H₅Br₂N
MW: 250.92 g/mol
MDL Number: MFCD00234955
CAS Number: 3430-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR