2-(Trifluoromethyl)benzenesulfonyl chloride ≥97%

Synonyms: 2-(Trifluoromethyl)benzene-1-sulphonyl chloride

491624-10G
220071-926EA 120.62 USD
220071-926
2-(Trifluoromethyl)benzenesulfonyl chloride ≥97%
2-(Trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Boiling Pt: 123 - 126 °C (1013 hPa)
MDL Number: MFCD00051696
CAS Number: 776-04-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR