6-Methylchromone hydrate ≥98%, Sigma-Aldrich®

Warning


M-Clarity™ quality level = MQ100

417475-1G
220063-638EA 62.46 USD
220063-638
6-Methylchromone hydrate ≥98%, Sigma-Aldrich®
6-Methylchromone hydrate

6-Methylchromone hydrate is a chromone derivative. Chromones are cyclic compounds containing nine carbon atoms with sp2 hybridization. They are commonly referred as 1,4-benzapyranes or γ-benzopyrones.


Application
6-Methylchromone may be employed as activated alkene in the methanolic trimethylamine or sodium methoxide catalyzed Baylis-Hillman coupling reaction with aromatic and aliphatic aldehydes.

Formula: C₁₀H₈O₂·xH₂O
MW: 160.17 g/mol (anhy)
MDL Number: MFCD00209598
CAS Number: 207511-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR