Dibenzyl sulfide ≥95.0%

33820-100G-F
220057-645EA 112 USD
220057-645
Dibenzyl sulfide ≥95.0%
Dibenzyl sulphide
Formula: C₁₄H₁₄S
MW: 214.33 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 47 - 50 °C
Density: 1.0712 g/cm³ (50 °C)
MDL Number: MFCD00004784
CAS Number: 538-74-9
EINECS: 208-703-6
UN: 3335
ADR: 9,
Merck Index: 13,01147

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR