2-Fluoro-6-nitrotoluene ≥98%

247685-10G
220047-891EA 466.15 USD
220047-891
2-Fluoro-6-nitrotoluene ≥98%
2-Fluoro-6-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1.2766 g/cm³ (20 °C)
MDL Number: MFCD00007197
CAS Number: 769-10-8
EINECS: 212-203-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR