1-Methoxynaphthalene

Synonyms: Methyl 1-naphthyl ether

154571-50G
220038-203EA 214.31 USD
220038-203
1-Methoxynaphthalene
1-Methoxynaphthalene
Formula: C₁₁H₁₀O
MW: 158.2 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 5 °C
Density: 1.09 g/cm³ (20 °C)
MDL Number: MFCD00003924
CAS Number: 2216-69-5
EINECS: 218-696-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR