VWR® Cheesecloth Wipers
Supplier: VWR International
21910-105 21910-107 46610-054
21910-105PK 109.83 USD
21910-105 21910-107 46610-054
VWR® Cheesecloth Wipers
Wipes Reusable Cleaning Cloths Cloths for Delicate Tasks
Use to wipe, clean, or dry cuvettes, delicate glassware, syringes, pipets, and pH electrodes.
  • Available in convenient dispenser box or on economical bolt
  • Inert to even harsh solvents
  • Mesh size: grade 10
Cheesecloth wipers are ideal for filtering and cleaning
Wipers meet cleanliness specifications of USP and Federal Specification CCC-G-101c.
Bleached, all-purpose cheesecloth wipers are brilliant white and certified 100% pure cotton of reagent-grade quality and purity. They feature a unique combination of cleanliness, softness, absorbency, andMore Product Information