VWR® Traceable® Minimum/Maximum Memory Thermometers
Supplier: VWR International
21800-076 62379-549 89094-786
21800-076EA 93.38 USD
21800-076 62379-549 89094-786
VWR® Traceable® Minimum/Maximum Memory Thermometers
Thermometers Digital Thermometers
Thermometers are ideal for monitoring solutions, refrigerators, and room temperature.
  • Traceable
  • High-impact ABS housing
  • Fits in a vaccine tray
  • Highly visible LCD digits
The 1 cm (3/8") high LCD digits display current, minimum, and maximum of ambient or probe temperature with the exact time and date when they occurred. Sensor with 182.9 cm (6') cable performs accurately even when both are under water. Features high-impact ABS housing. Thermometer with special bottle probe is specifically designed for refrigerator and freezer applications. VaccineMore Product Information