Nalgene® Top Works™ Aseptic Closure System, Silicone, for Bottles and Carboys, Thermo Scientific
Supplier: Thermo Fisher Scientific
2135-5303 2135-8302 2135-8303 2135-3803 2135-5300 2135-5302 2135-8300 2132-1003
21020-210CS 461.67 USD
21020-210 21020-214 21020-216 21020-204 21020-206 21020-208 21020-212 21020-254
Nalgene® Top Works™ Aseptic Closure System, Silicone, for Bottles and Carboys, Thermo Scientific
Closures Bottle Closures
Provides an aseptic seal for transferring high-purity fluids
Eight configurations permit precise matching of Top Works components to bottles and carboys. Polypropylene closures have center holes. They are modified to clamp a silicone stopper onto the bottle or carboy, creating a primary elastomeric seal. High-purity, platinum-cured silicone tubing ensures the fluid contact areas are crevice-free, which helps eliminate cross-contamination.
Materials are food-grade and noncytotoxic. Complies with U.S.P. Class VI requirements. Autoclavable.
For use with 125 mL to 50 L plastic bottlesMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable