Nalgene® Wide Mouth Centrifuge Bottle with Sealing Cap, Polycarbonate, Conical-Bottom, Thermo Scientific
Supplier: Thermo Fisher Scientific
3144-0175
21010-608CS 1544.64 USD
21010-608
Nalgene® Wide Mouth Centrifuge Bottle with Sealing Cap, Polycarbonate, Conical-Bottom, Thermo Scientific
Bottles Centrifuge Bottles
This transparent centrifuge bottle is manufactured from polycarbonate and is ideal for cell culture and protein biochemistry applications
Note: When autoclaving, place cap on top of container without engaging threads. Bottles must be used with adapter 21020-249. For proper performance, fill bottle to at least 80% of the total capacity.
It is graduated in 25 ml increments. Leakproof and autoclavable.
Información para pedidos: Supplied with polypropylene sealing cap with retained silicone gasket.