Eppendorf® Safe-Lock Tubes, 2.0 ml - Microtubes
Supplier: Eppendorf
022363379 022363395 022363417 022363476 022363344 022363352 022363433 022363450 022600044 0030123620
20901-541CS 113.63 USD
20901-541 20901-542 20901-543 20901-546 62111-754 20901-540 20901-544 20901-545 53511-998 76019-016
Eppendorf® Safe-Lock Tubes, 2.0 ml - Microtubes
Tubes Microcentrifuge Tubes
Eppendorf® Safe-Lock 2.0 ml microtube ensure secure sample preservation. With an innovative Safe-Lock mechanism, these microtubes prevent accidental openings, providing reliability in a 2.0 ml capacity. Ideal for various applications, they combine durability, ease of use, and sample protection.
  • Safe-Lock tube with a hinged lid prevents accidental opening during incubation and storage
  • g-Safe® technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30000×g
  • g-Safe® technology prevents
More Product Information