Threadlocker 262™ Permanent-Grade Adhesive, Loctite®, Henkel
Supplier: Loctite
135374
19807-518CS 640.56 USD
19807-518
Threadlocker 262™ Permanent-Grade Adhesive, Loctite®, Henkel
Adhesives
Permanent, high-strength product for locking fasteners 6.4 to 19mm (¼ to ¾") in diameter that must withstand heavy shock and vibration.
  • Permanent-grade
  • High-strength
  • Prevents rust and corrosion in extreme chemical and environmental conditions
Localized heating and hand tools are required to separate parts. Solvents will not weaken the adhesive bond.
Certificaciones: Meets Mil-Spec (S-46163A) for existing designs, NSF P1, CFIA, and American Bureau of Shipping standards.