VWR® Premium Tissue Cassettes
Supplier: VWR International
18000-000 18000-002 18000-004 18000-006 18000-008 18000-010 18000-012 18000-016 18000-018 18000-020
18000-000PK 419.93 USD
18000-000 18000-002 18000-004 18000-006 18000-008 18000-010 18000-012 18000-016 18000-018 18000-020
VWR® Premium Tissue Cassettes
Embedding Cassettes Tissue Processing Cassettes
These premium cassettes feature a fluidic flow design for the optimization of tissue processing and embedding.
  • Thin cassette design
  • Reagent-resistant acetyl polymer resin mold
  • Vented side walls maximize fluid exchange
Unique design has vented side walls to maximize fluid exchange
Thin cassettes have a similar construction but are 50% thinner than standard cassettes. This innovative design maximizes processing efficiency by doubling the number of cassettes that can be processed at once and by halving the storage area required for paraffin blocks.More Product Information