Nalgene® Air-Tight Pails, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
7102-0080 7102-0140
17308-022CS 775.44 USD
17308-022 17308-044
Nalgene® Air-Tight Pails, Low-Density Polyethylene, Thermo Scientific
Buckets
Close-fitting, airtight covers make these pails useful for specimens, storage and general utility
LDPE construction is suitable for use with formalin. Handle is a nylon-coated wire bail. Lids are white.
Certificaciones: Meets OSHA Standard 29 CFR 1910.1030 for use as protection against bloodborne pathogens.
Información para pedidos: Comes with lids and handles.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental Certification ISO14001 (Certificate Registration No. EMS 752603)