VWR® Micro Centrifuge Tube with Flat Screw-Cap
Supplier: VWR International
16466-030 16466-032 16466-034 16466-036 16466-038 16466-040 16466-042 16466-044 16466-046 16466-048 16466-050 16466-052 16466-054 16466-056 16466-058 16466-060 16466-062 16466-064
16466-030CS 698.51 USD
16466-030 16466-032 16466-034 16466-036 16466-038 16466-040 16466-042 16466-044 16466-046 16466-048 16466-050 16466-052 16466-054 16466-056 16466-058 16466-060 16466-062 16466-064
VWR® Micro Centrifuge Tube with Flat Screw-Cap
Tubes Microcentrifuge Tubes
These tubes are made from ultra-clear USP Class VI PP that allows easy viewing of contents.
  • RNase-, DNase-, and endotoxin-free
  • Engraved graduation ensure accuracy
  • Fit most common rotors
  • Smooth surface for applying labels
  • Sterile tubes are sterilized by irradiation
They feature universal screw threads for use with most popular brand screw caps. Low-profile caps include a silicone O-ring, ensuring a secure seal. Please kindly note: We began to use silicone O-ring from May 1st, 2022 and stopped manufacturing the MCT cap with the ethylene propylene O-ringMore Product Information