Emergency Eye Wash Station, Dynalon
Supplier: Dynalon
205165
16354-860PK 461.24 USD
16354-860
Emergency Eye Wash Station, Dynalon
First Aid Supplies Eyewash Stations and Eyewash Bottles
First EYD station with integral eye wash bottle.
  • A drain tube releases used fluid
  • Maximum comfort and ease of use
  • Capacity of bottle is 500 ml (16 oz.)
This station is molded of high-impact green polystyrene complete with polyethylene eye wash bottle. The unique bottle is designed to allow quick and immediate flushing to remove foreign matters and help prevent serious eye damage. The head has an air-vent valve to assure air refill of free space in the bottle thus preventing drawback of contaminated wash into the bottle. A perforated spray plate in theMore Product Information