Nalgene® Probe Adapter Closure, PP, Thermo Scientific
Supplier: Thermo Fisher Scientific
2145-0384
16225-250CS 264.82 USD
16225-250
Nalgene® Probe Adapter Closure, PP, Thermo Scientific
Closures Bottle Closures
Nalgene Probe Adapter Closures allow for insertion of 7- to 14mm diameter probes into Thermo Scientific Nalgene Culture Vessels and Bottles with 38-430 neck finishes
The probe adapter proves a seal between the external environment and the interior of the culture vessels to prevent contamination. The adapter includes a silicone gasket to ensure a leak-proof seal.
Order Now
SPECIFICATIONS
Environmentally Preferable