Straight Sided Rounds, Wide Mouth, Qorpak®
Supplier: Qorpak
GLC-07451 GLC-07897 GLC-01664 GLC-01605 GLC-01684 GLC-01628 GLC-01732 GLC-01596 GLC-01595 GLC-01624 GLC-01681 GLC-01705 GLC-01586 GLC-01611 GLC-01634 GLC-01672 GLC-01692 GLC-01721 GLC-01592 GLC-01620 GLC-01642 GLC-01677 GLC-01700 GLC-01728 GLA-00861 GLA-00864 GLA-00857 GLA-00850 GLA-00859 GLA-00867 GLC-01584 GLC-01629 GLC-01666 GLC-01635 GLC-01673 GLC-01693 GLC-01665 GLC-01717 GLC-04910 GLA-00854 239228 250239 239223 239224 239225 239227 239229 239230 250240 272175 272174 272173 272172 272171 272170 GLC-01581 GLC-01606 GLC-01685
11311-142CS 83.56 USD
11311-142 11311-148 11311-386 11311-388 11311-390 11311-662 14002-387 16054-140 16054-142 16054-146 16054-148 16054-176 16054-182 16081-040 16087-308 16151-580 16194-100 16194-518 16195-022 16195-033 16195-044 16195-055 16195-059 16195-061 16195-063 16195-088 16195-102 16195-306 16195-328 16195-340 16195-361 16195-383 16195-408 16195-667 16195-689 16195-703 16195-725 16195-727 16195-747 16195-769 16195-770 16195-771 16195-772 16218-176 16218-178 16218-180 16218-182 36319-103 36319-111 36319-113 36319-145 36319-147 36319-149 36319-151 36319-231 36319-232 36319-237 82024-648 82027-766 89204-942 10847-230 10847-280 76170-386 76170-396 76170-400 76170-402 76170-404 76170-406 76170-408 76170-410 76170-422 76419-794 76419-796 76419-798 76419-800 76419-802 76419-804 76487-202 76487-204 76487-206
Straight Sided Rounds, Wide Mouth, Qorpak®
Jars General Use Jars
Clear, flint glass straight sided round jars are designed without shoulders for maximum storage capacity and content visibility.
  • Made from Type III Soda Lime Glass
  • USP certified Pharma grade glass
  • Clear glass is appropriate when visibility is important and protection from UV rays is not necessary
The wide mouth opening is ideal for mixing, storing, and sampling. Many closures are available to suit particular needs.
Información para pedidos: Bottles 36319-113, 36319-145, 36319-147, 36319-149, 36319-151, and 16151-580 are supplied pre-cleaned forMore Product Information