WHEATON® Polypropylene Screw Cap, Linerless, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
239501 239501-02 239501-03 239501-06 239506 239506-02 239506-03 239506-04 239506-06 239510 239510-02 239510-03 239510-04 239510-06 239512 239512-02 239512-04 239516 239516-02 239516-03 239516-04
16153-463CS 71.99 USD
16153-463 16153-465 16153-467 16153-471 16153-473 16153-475 16153-477 16153-479 16153-481 16153-483 16153-485 16153-487 16153-489 16153-491 16153-493 16153-495 16153-499 16153-503 16153-505 16153-507 16153-509
WHEATON® Polypropylene Screw Cap, Linerless, DWK Life Sciences
Closures Bottle Closures
A variety of polypropylene caps are offered separately for Wheaton Leak Resistant Narrow Mouth Bottles
Caps can be autoclaved for 15 minutes at 121ºC at 15 psi. Polypropylene caps are available in natural, white, red, blue and green
Order Now
SPECIFICATIONS
Environmentally Preferable