KIMAX® Nessler Color Comparison Tubes, Tall Form, APHA Standard, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
45315A-100 45315-100 45315B-50 45315B-100 45325B-50100 45315-50 45315-50100 45325A-50 45315A-50100 45315A-50
16143-254EA 409.44 USD
16143-254 66176-326 66176-507 66176-529 66176-686 66176-305 66176-348 66176-620 72640-048 16143-253
KIMAX® Nessler Color Comparison Tubes, Tall Form, APHA Standard, Kimble Chase
Tubes Nessler Tubes
These narrow, tall tubes with a long scale length feature 'Shadowless Bottoms' that transmit undistorted light from a source beneath tube. All graduation lines and legends are printed in white ceramic enamel.
  • Ideal for color comparison
  • Shadowless bottom of each tube transmits undistorted light
  • Rings and legend are durable white ceramic enamel
Available unmatched or in matched sets of 6 or 12.
The 50 to 100 ml tubes with cap stopper were designed from ASTM D1209. [ST] 24/12 taper cap style stoppers need not be removed when making readings. The word 'More Product Information