Nalgene® Large Square Bottle, Polypropylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2122-0010
16130-005CS 273.6 USD
16130-005
Nalgene® Large Square Bottle, Polypropylene, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Space-saving bottle features convenient, molded-in handgrips.
  • Excellent for storing and sterilizing contaminated slides and other small lab items
  • Not leakproof
  • Autoclavable
  • Cap size: 100-415
Note: Before autoclaving, set cap on top of the bottle without engaging threads.
Información para pedidos: Supplied with polypropylene screw caps assembled on bottles.
Order Now
SPECIFICATIONS
Environmentally Preferable