SP Bel-Art Scrapers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36831-0000 F36832-0000 F36831-0001 F36832-0001 F36830-0001 F36830-0000
13197-242EA 252.34 USD
13197-242 13197-244 16126-598 16126-600 47750-092 16126-594
SP Bel-Art Scrapers, Bel-Art Products, a part of SP
Scrapers
Smooth, chemical-resistant plastic scrapers are useful for removing residues from filters, cleaning containers, mixing and distributing solids in fluids, spreading pastes over large areas, and many other scraping, spreading, stirring or residue removing tasks.
  • Smooth, chemical-resistant surface
  • Available in LDPE or HDPE models
  • Used for a variety of applications
  • Chisel edge on one end, hexagonal shank on the other
The blade is 12.7 cm (5”) wide x 15.2 cm (6”) long and tapers from 9.5 mm (⅜”) down to a chisel edge. The upper end extends into a hexagonalMore Product Information