Corning® Polypropylene Snap-Seal Sample Containers, Corning
Supplier: Corning
1730-2C 1730-4L 1730-10 1730-4H 1730-8
16126-010CS 227.01 USD
16126-010 16126-014 16126-022 16126-030 16126-018 16126-026
Corning® Polypropylene Snap-Seal Sample Containers, Corning
Sample Containers General Purpose Containers
PP, disposable, translucent.
  • Snap-Seal locking device keeps the cap closed and secure
  • Hinged cap reduces the chance of sample contamination
  • Graduations in both millilitres and ounces
  • Rough source for marking
These wide mouth sample containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly.
Order Now
SPECIFICATIONS
Environmentally Preferable