Rosetta-gami™ 2 Competent Cells, Expression Strains, MilliporeSigma
Supplier: MilliporeSigma
71350-3 71352-4 71350-4 71351-4 71352-3 71351-3
16003-952EA 223.82 USD
16003-952 16003-962 16003-954 16003-958 16003-960 16003-956
Rosetta-gami™ 2 Competent Cells, Expression Strains, MilliporeSigma
Competent Cells
Rosetta-gami™ 2 host strains combine the advantages of Rosetta 2 and Origami 2 strains to alleviate codon bias and enhance disulfide bond formation in the cytoplasm when heterologoous proteins are expressed in E. coli.
  • Guaranteed efficiency: 2×10⁶ cfu/µg
These trxB/gor mutants are compatible with kanamycin-resistant vectors, and carry the chloramphenicol-resistant pRARE2 plasmid, which supplies seven rare tRNAs.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNAMore Product Information