Evacuable Powdered Sample Briquetting Dies, Chemplex®
Supplier: Chemplex
4038 4035 4037 4045 4050 4055 4060 4013
15566-200EA 5055.46 USD
15566-200 15566-202 15566-206 15566-208 15566-210 15566-212 15566-214 15566-216 89079-864 89079-866 89079-868 89079-870
Evacuable Powdered Sample Briquetting Dies, Chemplex®
Presses
For producing firm, durable IR or XRF powdered sample briquettes.
Tungsten carbide pellets are optionally available for briquetting very hard or abrasive powdered sample materials.
Precision-ground stainless steel. Each die set includes a main body with provision for attaching a vacuum line, a base with O-ring, micro-polished stainless steel pellets, a plunger with O-ring, and a sample extractor cup to facilitate sample removal.
Order Now
SPECIFICATIONS
Environmentally Preferable