XRF Sample Cup Series 1500, Chemplex®
Supplier: Chemplex
1530 1540
15566-168PK 135.91 USD
15566-168 15566-170
XRF Sample Cup Series 1500, Chemplex®
Sample Containers XRF Sample Cups
Polyethylene sample cups designed with double open ends.
Adaptable to contain many types of sample materials in air, inert gas and evacuated conditions when used with microporous film and vented sample cup caps. Can be top sample loaded after preattaching thin-film sample supports. Consists of a cell and two identical snap-on rings. Cup caps and film sold separately. Height: 23mm (0.9")
Order Now
SPECIFICATIONS
Environmentally Preferable