SpectroBlend® additive, powder, Chemplex® 44 µm
Supplier: CHEMPLEX INDUSTRIES
660
15566-078EA 371.26 USD
15566-078
SpectroBlend® additive, powder, Chemplex® 44 µm
SpectroBlend® additive
A 44µm powder well suited for blending and as a diluent for high concentrations. Well balanced with abrasive and lubricious constituents for grinding most sample materials emphasizing difficult-to-process samples requiring long grinding cycles in metallic comminution vials or dishes. Resistant to degradation by thermal and irradiation exposure. Samples have “plasticized” surfaces. Available in bulk powder and ½ gm pre-weighed tablets for ease of dispensing.
  • Improve powdered sample preparations
  • Reduce particles to uniform size and distribution
  • Promote sample homogeneity
  • Transform powders into “Plasticized” pellets
  • Resolve sample pellet chipping or flaking
Chemical Composition:
C: 81.0%; O: 2.9%; H: 13.5%; N: 2.6%