Pure-Touch Pumps, Menda
Supplier: Desco
35500 35501 35502 35503
15431-766EA 20.82 USD
15431-766 15431-768 15431-770 15431-772
Pure-Touch Pumps, Menda
Bottle Top Dispensers
The pure-touch pump features a purity protector to stop fluid from re-entering the bottle.
  • Neck size: 38-400 thread
  • One-handed operation frees operator's hands
  • Precision machined valves for smooth and consistent operation
  • Cam-lock hinge keeps lid open and closed, reduces odors
  • Dispenses approximately 0.2 cc of liquid with each pump
Pumps feature no silicone or latex, and are therefore perfect for ultra clean environments. Ideal for cleaning with cotton balls, pads, and brushes. Fits all standard Menda HDPE, glass, and steel bottles. Purity protectorMore Product Information